4-Brom-N-tert-butylbenzamid
Lieferant: COMBI-BLOCKS
Synonyme:
4-Bromo-N-(tert-butyl)benzamide
CA-4396-5G
CA-4396-25G
CA-4396-1G
COBBCA-4396-5GEA
451
EUR
COBBCA-4396-5G
COBBCA-4396-25G
COBBCA-4396-1G
4-Brom-N-tert-butylbenzamid
4-Brom-N-tert-butylbenzamid
Formel:
C₁₁H₁₄BrNO Molecular Weight: 256.1400 g/mol |
MDL:
MFCD00462539 CAS-Nummer: 42498-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...