4-Brom-N-butylbenzamid

Lieferant: COMBI-BLOCKS

CA-4421-5G CA-4421-25G CA-4421-1G
COBBCA-4421-5GEA 437 EUR
COBBCA-4421-5G COBBCA-4421-25G COBBCA-4421-1G
4-Brom-N-butylbenzamid
4-Brom-N-butylbenzamid
Formel: C₁₁H₁₄BrNO
MDL: MFCD00456169
CAS-Nummer: 78080-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR