3-Brom-N-methyl-5-nitrobenzamid

Lieferant: COMBI-BLOCKS

CA-4381-1G CA-4381-5G
COBBCA-4381-1GEA 273 EUR
COBBCA-4381-1G COBBCA-4381-5G
3-Brom-N-methyl-5-nitrobenzamid
3-Brom-N-methyl-5-nitrobenzamid
Formel: C₈H₇BrN₂O₃
MDL: MFCD09475916
CAS-Nummer: 90050-52-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR