4-Brom-2-methoxy-N-methylbenzamid

Lieferant: COMBI-BLOCKS

CA-4994-5G CA-4994-1G
COBBCA-4994-5GEA 612 EUR
COBBCA-4994-5G COBBCA-4994-1G
4-Brom-2-methoxy-N-methylbenzamid
4-Brom-2-methoxy-N-methylbenzamid
Formel: C₉H₁₀BrNO₂
MDL: MFCD16618946
CAS-Nummer: 1228957-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR