4-Brom-3-methoxy-N-propylbenzamid

Lieferant: COMBI-BLOCKS

CA-4649-5G CA-4649-1G
COBBCA-4649-5GEA 451 EUR
COBBCA-4649-5G COBBCA-4649-1G
4-Brom-3-methoxy-N-propylbenzamid
4-Brom-3-methoxy-N-propylbenzamid
Formel: C₁₁H₁₄BrNO₂
MDL: MFCD11504873
CAS-Nummer: 1072944-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR