3-Brom-N-(2-hydroxyethyl)benzamid

Lieferant: COMBI-BLOCKS

CA-4482-5G CA-4482-1G
COBBCA-4482-5GEA 884 EUR
COBBCA-4482-5G COBBCA-4482-1G
3-Brom-N-(2-hydroxyethyl)benzamid
3-Brom-N-(2-hydroxyethyl)benzamid
Formel: C₉H₁₀BrNO₂
MDL: MFCD01249209
CAS-Nummer: 57728-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR