4-Brom-3-methoxy-N-phenylbenzamid

Lieferant: COMBI-BLOCKS

CA-4644-5G CA-4644-1G
COBBCA-4644-5GEA 303 EUR
COBBCA-4644-5G COBBCA-4644-1G
4-Brom-3-methoxy-N-phenylbenzamid
4-Brom-3-methoxy-N-phenylbenzamid
Formel: C₁₄H₁₂BrNO₂
MDL: MFCD11504868
CAS-Nummer: 1072944-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR