Methyl-5-brom-2-(methylthio)benzoat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 5-bromo-2-(methylthio)benzoate
CA-4245-1G
CA-4245-5G
CA-4245-25G
COBBCA-4245-1GEA
163
EUR
COBBCA-4245-1G
COBBCA-4245-5G
COBBCA-4245-25G
Methyl-5-brom-2-(methylthio)benzoat
Methyl-5-brom-2-(methylthio)benzoat
Formel:
C₉H₉BrO₂S Molecular Weight: 261,1356 g/mol |
MDL:
MFCD09258758 CAS-Nummer: 929000-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...