4-Brom-N,N-dimethyl-2-fluorbenzamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-2-fluoro-N,N-dimethylbenzamide

CA-4504-5G CA-4504-1G
COBBCA-4504-5GEA 451 EUR
COBBCA-4504-5G COBBCA-4504-1G
4-Brom-N,N-dimethyl-2-fluorbenzamid
4-Brom-N,N-dimethyl-2-fluorbenzamid
Formel: C₉H₉BrFNO
Molecular Weight: 246,0763 g/mol
MDL: MFCD09878360
CAS-Nummer: 749927-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR