2-Brom-N-butyl-5-fluorbenzamid

Lieferant: COMBI-BLOCKS

CA-4448-1G CA-4448-5G
COBBCA-4448-1GEA 149 EUR
COBBCA-4448-1G COBBCA-4448-5G
2-Brom-N-butyl-5-fluorbenzamid
2-Brom-N-butyl-5-fluorbenzamid
Formel: C₁₁H₁₃BrFNO
MDL: MFCD09800917
CAS-Nummer: 951884-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR