2-Brom-5-fluor-N-methylbenzamid

Lieferant: COMBI-BLOCKS

CA-4393-5G CA-4393-1G
COBBCA-4393-5GEA 451 EUR
COBBCA-4393-5G COBBCA-4393-1G
2-Brom-5-fluor-N-methylbenzamid
2-Brom-5-fluor-N-methylbenzamid
Formel: C₈H₇BrFNO
MDL: MFCD09475889
CAS-Nummer: 171426-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR