2-Brom-5-fluor-N-propylbenzamid
Lieferant: COMBI-BLOCKS
CA-4435-5G
CA-4435-1G
COBBCA-4435-5GEA
1130
EUR
COBBCA-4435-5G
COBBCA-4435-1G
2-Brom-5-fluor-N-propylbenzamid
2-Brom-5-fluor-N-propylbenzamid
Formel:
C₁₀H₁₁BrFNO |
MDL:
MFCD09800906 CAS-Nummer: 951884-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...