3-Brom-4-fluor-N-propylbenzamid

Lieferant: COMBI-BLOCKS

CA-4608-1G CA-4608-25G CA-4608-5G
COBBCA-4608-1GEA 74.2 EUR
COBBCA-4608-1G COBBCA-4608-25G COBBCA-4608-5G
3-Brom-4-fluor-N-propylbenzamid
3-Brom-4-fluor-N-propylbenzamid
Formel: C₁₀H₁₁BrFNO
MDL: MFCD11053797
CAS-Nummer: 1065074-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR