4-Brom-2-fluor-N-propylbenzamid

Lieferant: COMBI-BLOCKS

CA-4998-5G CA-4998-1G
COBBCA-4998-5GEA 451 EUR
COBBCA-4998-5G COBBCA-4998-1G
4-Brom-2-fluor-N-propylbenzamid
4-Brom-2-fluor-N-propylbenzamid
Formel: C₁₀H₁₁BrFNO
MDL: MFCD09935765
CAS-Nummer: 1016811-41-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR