2-Brom-5-fluor-N-(2-hydroxyethyl)benzamid

Lieferant: COMBI-BLOCKS

CA-4443-1G CA-4443-5G
COBBCA-4443-1GEA 149 EUR
COBBCA-4443-1G COBBCA-4443-5G
2-Brom-5-fluor-N-(2-hydroxyethyl)benzamid
2-Brom-5-fluor-N-(2-hydroxyethyl)benzamid
Formel: C₉H₉BrFNO₂
MDL: MFCD09800913
CAS-Nummer: 951884-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR