1-Brom-5-fluor-2,4-dinitrobenzol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromo-2,4-dinitro-5-fluorobenzene

HC-6682-5G HC-6682-25G HC-6682-1G
COBBHC-6682-5GEA 141 EUR
COBBHC-6682-5G COBBHC-6682-25G COBBHC-6682-1G
1-Brom-5-fluor-2,4-dinitrobenzol
1-Brom-5-fluor-2,4-dinitrobenzol
Formel: C₆H₂BrFN₂O₄
MDL: MFCD00024181
CAS-Nummer: 400-91-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR