2-Brom-N,N-diethyl-4-(trifluormethoxy)benzolsulfonamid

Lieferant: COMBI-BLOCKS

HC-6409-1G HC-6409-5G
COBBHC-6409-1GEA 149 EUR
COBBHC-6409-1G COBBHC-6409-5G
2-Brom-N,N-diethyl-4-(trifluormethoxy)benzolsulfonamid
2-Brom-N,N-diethyl-4-(trifluormethoxy)benzolsulfonamid
Formel: C₁₁H₁₃BrF₃NO₃S
MDL: MFCD09878396
CAS-Nummer: 957062-75-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR