2-Brom-N-butyl-4-(trifluormethoxy)benzolsulfonamid
Lieferant: COMBI-BLOCKS
HC-6408-5G
HC-6408-1G
COBBHC-6408-5GEA
0
EUR
COBBHC-6408-5G
COBBHC-6408-1G
2-Brom-N-butyl-4-(trifluormethoxy)benzolsulfonamid
2-Brom-N-butyl-4-(trifluormethoxy)benzolsulfonamid
Formel:
C₁₁H₁₃BrF₃NO₃S |
MDL:
MFCD09878395 CAS-Nummer: 957062-76-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...