3-Brom-4-fluorthiophenol

Lieferant: FLUOROCHEM

033393-1G 033393-5G
FLUO033393-1GEA 351 EUR
FLUO033393-1G FLUO033393-5G
3-Brom-4-fluorthiophenol
3-Brom-4-fluorthiophenol
Formel: C₆H₄BrFS
MDL: MFCD09475474
CAS-Nummer: 942473-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR