Methyl-5-brom-2-iodbenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 5-bromo-2-iodobenzoate
CA-4979-5G
CA-4979-25G
CA-4979-1G
COBBCA-4979-5GEA
102
EUR
COBBCA-4979-5G
COBBCA-4979-25G
COBBCA-4979-1G
Methyl-5-brom-2-iodbenzoat
Methyl-5-brom-2-iodbenzoat
Formel:
C₈H₆BrIO₂ Molecular Weight: 340,9400 g/mol |
MDL:
MFCD00144771 CAS-Nummer: 181765-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...