1-Brom-2-iod-5-nitrobenzol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-4-iodonitrobenzene

HC-3045-5G HC-3045-1G
COBBHC-3045-5GEA 465 EUR
COBBHC-3045-5G COBBHC-3045-1G
1-Brom-2-iod-5-nitrobenzol
1-Bromo-2-iodo-5-nitrobenzene
Formel: C₆H₃BrINO₂
Molecular Weight: 327,9000 g/mol
MDL: MFCD09878135
CAS-Nummer: 7149-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR