N-(2-Aminoethyl)-4-brombenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0093-5G OT-0093-1G
COBBOT-0093-5GEA 912 EUR
COBBOT-0093-5G COBBOT-0093-1G
N-(2-Aminoethyl)-4-brombenzolsulfonamid
N-(2-Aminoethyl)-4-brombenzolsulfonamid
Formel: C₈H₁₁BrN₂O₂S
MDL: MFCD06659916
CAS-Nummer: 90002-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR