1-Brom-4-(methylsulfonyl)-2-nitrobenzol

Lieferant: COMBI-BLOCKS

CS-9011-1G CS-9011-25G CS-9011-5G
COBBCS-9011-1GEA 104 EUR
COBBCS-9011-1G COBBCS-9011-25G COBBCS-9011-5G
1-Brom-4-(methylsulfonyl)-2-nitrobenzol
1-Bromo-4-(methylsulphonyl)-2-nitrobenzene
Formel: C₇H₆BrNO₄S
Molecular Weight: 280,0958 g/mol
MDL: MFCD00054605
CAS-Nummer: 94832-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR