5-Brom-N,N-diethyl-2-methoxybenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0198-5G OT-0198-1G
COBBOT-0198-5GEA 451 EUR
COBBOT-0198-5G COBBOT-0198-1G
5-Brom-N,N-diethyl-2-methoxybenzolsulfonamid
5-Brom-N,N-diethyl-2-methoxybenzolsulfonamid
Formel: C₁₁H₁₆BrNO₃S
MDL: MFCD02919424
CAS-Nummer: 428471-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR