5-Brom-2-methoxybenzolsulfonamid
Lieferant: COMBI-BLOCKS
OT-0310-5G
OT-0310-1G
COBBOT-0310-5GEA
692
EUR
COBBOT-0310-5G
COBBOT-0310-1G
5-Brom-2-methoxybenzolsulfonamid
5-Brom-2-methoxybenzolsulfonamid
Formel:
C₇H₈BrNO₃S |
MDL:
MFCD06808831 CAS-Nummer: 23095-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...