3-Brom-N-(2-hydroxyethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0147-5G OT-0147-25G OT-0147-1G
COBBOT-0147-5GEA 0 EUR
COBBOT-0147-5G COBBOT-0147-25G COBBOT-0147-1G
3-Brom-N-(2-hydroxyethyl)benzolsulfonamid
3-Brom-N-(2-hydroxyethyl)benzolsulfonamid
Formel: C₈H₁₀BrNO₃S
MDL: MFCD08436008
CAS-Nummer: 911111-96-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR