4-Brom-N-(2-(TBDMSO)ethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-N-[2-(tert-butyldimethylsilyloxy)ethyl]benzenesulphonamide, 4-Bromo-N-(2-((tert-butyldimethylsilyl)oxy)ethyl)benzenesulphonamide

CS-9086-1G CS-9086-5G
COBBCS-9086-1GEA 0 EUR
COBBCS-9086-1G COBBCS-9086-5G
4-Brom-N-(2-(TBDMSO)ethyl)benzolsulfonamid
4-Brom-N-(2-(TBDMSO)ethyl)benzolsulfonamid
Formel: C₁₄H₂₄BrNO₃SSi
Molecular Weight: 394.400 g/mol
MDL: MFCD06659896
CAS-Nummer: 850429-52-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR