3-Chlor-5-(diethylcarbamoyl)phenylboronsäure

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-5-(diethylcarbonyl)phenylboronic acid, (3-Chloro-5-(diethylcarbamoyl)phenyl)boronic acid

OR11644-1G OR11644-5G
APOSOR11644-1GEA 120 EUR
APOSOR11644-1G APOSOR11644-5G
3-Chlor-5-(diethylcarbamoyl)phenylboronsäure
3-Chlor-5-(diethylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₅BClNO₃
Molecular Weight: 255.5100 g/mol
MDL: MFCD09800862
CAS-Nummer: 957120-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR