3-Chlor-5-(diethylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Chloro-5-(diethylcarbonyl)phenylboronic acid, (3-Chloro-5-(diethylcarbamoyl)phenyl)boronic acid
BB-3248-5G
BB-3248-1G
COBBBB-3248-5GEA
1310
EUR
COBBBB-3248-5G
COBBBB-3248-1G
3-Chlor-5-(diethylcarbamoyl)phenylboronsäure
3-Chlor-5-(diethylcarbamoyl)phenylboronsäure
Formel:
C₁₁H₁₅BClNO₃ Molecular Weight: 255.5100 g/mol |
MDL:
MFCD09800862 CAS-Nummer: 957120-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...