3-Chlor-5-(ethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-5-(ethylcarbamoyl)phenyl)boronic acid

BB-3237-1G BB-3237-5G
COBBBB-3237-1GEA 375 EUR
COBBBB-3237-1G COBBBB-3237-5G
3-Chlor-5-(ethylcarbamoyl)phenylboronsäure
3-Chlor-5-(ethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₁BClNO₃
Molecular Weight: 227.4500 g/mol
MDL: MFCD09800857
CAS-Nummer: 957120-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR