3-(2-Chlorethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-((2-Chloroethyl)carbamoyl)phenyl)boronic acid

BB-3154-5G BB-3154-1G
COBBBB-3154-5GEA 725 EUR
COBBBB-3154-5G COBBBB-3154-1G
3-(2-Chlorethylcarbamoyl)phenylboronsäure
3-(2-Chlorethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₁BClNO₃
Molecular Weight: 227.4500 g/mol
MDL: MFCD08436013
CAS-Nummer: 874288-12-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR