3-(3-Chlorpropylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-((3-Chloropropyl)carbamoyl)phenyl)boronic acid

BB-3152-5G BB-3152-1G
COBBBB-3152-5GEA 695 EUR
COBBBB-3152-5G COBBBB-3152-1G
3-(3-Chlorpropylcarbamoyl)phenylboronsäure
3-(3-Chlorpropylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₃BClNO₃
Molecular Weight: 241,4800 g/mol
MDL: MFCD08436014
CAS-Nummer: 874288-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR