4-(2-Chlorethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3104-5G BB-3104-1G
COBBBB-3104-5GEA 0 EUR
COBBBB-3104-5G COBBBB-3104-1G
4-(2-Chlorethylcarbamoyl)phenylboronsäure
4-(2-Chlorethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₁BClNO₃
MDL: MFCD08436019
CAS-Nummer: 874460-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR