2-Chlor-4-methoxy-5-(methoxycarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-3284-1G
BB-3284-250M
COBBBB-3284-1GEA
742
EUR
COBBBB-3284-1G
COBBBB-3284-250M
2-Chlor-4-methoxy-5-(methoxycarbonyl)phenylboronsäure
2-Chlor-4-methoxy-5-(methoxycarbonyl)phenylboronsäure
Formel:
C₉H₁₀BClO₅ |
MDL:
MFCD09878324 CAS-Nummer: 957066-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...