2-Chlor-N,N-diethylbenzamid

Lieferant: COMBI-BLOCKS

CA-4985-5G CA-4985-25G
COBBCA-4985-5GEA 149 EUR
COBBCA-4985-5G COBBCA-4985-25G
2-Chlor-N,N-diethylbenzamid
2-Chlor-N,N-diethylbenzamid
Formel: C₁₁H₁₄ClNO
MDL: MFCD00018262
CAS-Nummer: 10345-79-6
EINECS: 233-754-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR