4-Chlor-N-tert-butylbenzamid

Lieferant: COMBI-BLOCKS

CA-4775-5G CA-4775-25G CA-4775-1G
COBBCA-4775-5GEA 127 EUR
COBBCA-4775-5G COBBCA-4775-25G COBBCA-4775-1G
4-Chlor-N-tert-butylbenzamid
4-Chlor-N-tert-butylbenzamid
Formel: C₁₁H₁₄ClNO
MDL: MFCD00461169
CAS-Nummer: 42498-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR