4-(4-Chlorphenoxy)thiophenol

Lieferant: CHEMPUR

05-0034.500 05-0034.1
CHMP05-0034.500EA 0 EUR
CHMP05-0034.500 CHMP05-0034.1
4-(4-Chlorphenoxy)thiophenol
4-(4-Chlorphenoxy)thiophenol
Formel: C₁₂H₉ClOS
MDL: MFCD04114345
CAS-Nummer: 59621-76-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR