3-Cyanbenzamid

Lieferant: SIA ENAMINE

EN300-31182.10 EN300-31182.5G
ENAMEN300-31182.10EA 0 EUR
ENAMEN300-31182.10 ENAMEN300-31182.5G
3-Cyanbenzamid
3-Cyanbenzamid
Formel: C₈H₆N₂O
MDL: MFCD00017574
CAS-Nummer: 3441-01-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR