3,4-Diaminobenzonitril

Lieferant: Thermo Scientific

SPB07644.250 SPB07644.1
MAYBSPB07644.250EA 0 EUR
MAYBSPB07644.250 MAYBSPB07644.1
3,4-Diaminobenzonitril
3,4-Diaminobenzonitril
Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
MDL: MFCD00723901
CAS-Nummer: 17626-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR