3-Hydroxy-5-nitrobenzonitril
Lieferant: COMBI-BLOCKS
AN-1132-1G
AN-1132-5G
COBBAN-1132-1GEA
497
EUR
COBBAN-1132-1G
COBBAN-1132-5G
3-Hydroxy-5-nitrobenzonitril
3-Hydroxy-5-nitrobenzonitril
Formel:
C₇H₄N₂O₃ Molecular Weight: 164.1183 g/mol |
MDL:
MFCD09258662 CAS-Nummer: 929000-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...