4-Hydroxy-3-nitrobenzonitril ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H55597.03 H55597.06
H55597.03EA 38.6 EUR
H55597.03 H55597.06
4-Hydroxy-3-nitrobenzonitril ≥98%
4-Hydroxy-3-nitrobenzonitril

Purity: 98% CAS No.: 3272-08-0 Molecular Formula: C7H4N2O3 Formula Weight: 164.12 Melting Point: 146-148° Beilstein Reference: 2048724

Formel: C₇H₄N₂O₃
Molecular Weight: 164.12 g/mol
Schmelzpunkt: 146…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00070775
CAS-Nummer: 3272-08-0
EINECS: 221-899-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR