2,6-Dihydroxy-3-nitrobenzonitril
Lieferant: Thermo Scientific
CD07514.250
CD07514.1
MAYBCD07514.250EA
0
EUR
MAYBCD07514.250
MAYBCD07514.1
2,6-Dihydroxy-3-nitrobenzonitril
2,6-Dihydroxy-3-nitrobenzonitril
Formel:
C₇H₄N₂O₄ |
CAS-Nummer:
233585-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...