4-Amino-N,N-dimethylbenzamid ≥97%

Lieferant: Thermo Scientific
Achtung

H50364.03 H50364.06
H50364.03EA 179 EUR
H50364.03 H50364.06
4-Amino-N,N-dimethylbenzamid ≥97%
4-Amino-N,N-dimethylbenzamid

Purity: 97+% CAS No.: 6331-71-1 Molecular Formula: C9H12N2O Formula Weight: 164.21 Melting Point: 151-153° MDL No.: MFCD00523648

Formel: C₉H₁₂N₂O
Molecular Weight: 164,21 g/mol
Schmelzpunkt: 151…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00523648
CAS-Nummer: 6331-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR