Methyl-2-(methylthio)benzoat

Lieferant: COMBI-BLOCKS

CA-4225-1G CA-4225-25G CA-4225-5G
COBBCA-4225-1GEA 48.4 EUR
COBBCA-4225-1G COBBCA-4225-25G COBBCA-4225-5G
Methyl-2-(methylthio)benzoat
Methyl-2-(methylthio)benzoat
Formel: C₉H₁₀O₂S
Molecular Weight: 182,24 g/mol
MDL: MFCD00068039
CAS-Nummer: 3704-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR