Methyl-4-(methylthio)benzoat

Lieferant: CHEMPUR

Synonyme: Methyl 4-(methylthio)benzoate

RH01345.250 RH01345.1
CHMPRH01345.250EA 0 EUR
CHMPRH01345.250 MAYBRH01345.1
Methyl-4-(methylthio)benzoat
Methyl-4-(methylthio)benzoat
Formel: C₉H₁₀O₂S
Molecular Weight: 182,239 g/mol
MDL: MFCD01313010
CAS-Nummer: 3795-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR