Probenecid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-[(dipropylamino)sulphonyl]benzoic acid

B20010.88 B20010.09
B20010.88EA 44.4 EUR
B20010.88 B20010.09
Probenecid ≥98%
Probenecid

Purity: 98% CAS No.: 57-66-9 Molecular Formula: C13H19NO4S Formula Weight: 285.36 Physical Form: Powder Appearance: White Melting Point: 198-199° Merck Reference: 14,7754 MDL No.: MFCD00038402

Formel: C₁₃H₁₉NO₄S
Molecular Weight: 285,4 g/mol
Schmelzpunkt: 198…199 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038402
CAS-Nummer: 57-66-9
EINECS: 200-344-3
Merck Index: 14,7754

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR