2-Cyan-6-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Cyano-6-fluorophenyl)boronic acid

FA-1631-5G FA-1631-1G FA-1631-250M
COBBFA-1631-5GEA 1080 EUR
COBBFA-1631-5G COBBFA-1631-1G COBBFA-1631-250M
2-Cyan-6-fluorphenylboronsäure
2-Cyan-6-fluorphenylboronsäure
Formel: C₇H₅BFNO₂
Molecular Weight: 164.9295 g/mol
MDL: MFCD09033623
CAS-Nummer: 656235-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR