3-Cyan-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Cyano-2-fluorophenyl)boronic acid

BB-5008-5G BB-5008-1G
COBBBB-5008-5GEA 409 EUR
COBBBB-5008-5G COBBBB-5008-1G
3-Cyan-2-fluorphenylboronsäure
3-Cyan-2-fluorphenylboronsäure
Formel: C₇H₅BFNO₂
Molecular Weight: 164,9295 g/mol
MDL: MFCD07374984
CAS-Nummer: 957121-05-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR