3-Cyan-4-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Cyano-4-fluorobenzeneboronic acid

BB-2327-5G BB-2327-1G
COBBBB-2327-5GEA 236 EUR
COBBBB-2327-5G COBBBB-2327-1G
3-Cyan-4-fluorphenylboronsäure
3-Cyan-4-fluorphenylboronsäure
Formel: C₇H₅BFNO₂
Molecular Weight: 164,93 g/mol
MDL: MFCD03095130
CAS-Nummer: 214210-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR