5-Cyan-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Cyano-2-fluorobenzeneboronic acid

BB-2609-5G BB-2609-1G
COBBBB-2609-5GEA 273 EUR
COBBBB-2609-5G COBBBB-2609-1G
5-Cyan-2-fluorphenylboronsäure
5-Cyan-2-fluorphenylboronsäure
Formel: C₇H₅BFNO₂
Molecular Weight: 164,9300 g/mol
MDL: MFCD03095147
CAS-Nummer: 468718-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR